Cole-Parmer Universal Pipette Tips with Filter, Low Retention, Sterile, 1000 µL; 10 Racks x 96 Tips
Representative image only.
Cole-Parmer Universal Pipette Tips with Filter, Low Retention, Sterile, 1000 µL; 10 Racks x 96 Tips
Cole-Parmer | Item: 7890133
Protect against cross-contamination with tips free of human touch
$260.05CAD / Case of 960
BUY MORE & SAVE
  • 1-9$260.05 CAD / Case of 960
  • 10+$234.05 CAD / Case of 960
$260.05CAD / Case of 960
BUY MORE & SAVE
  • 1-9$260.05 CAD / Case of 960
  • 10+$234.05 CAD / Case of 960
Product Description
Key Features
  • Pure porous polyethylene filter protects against cross-contamination
  • Durable polypropylene construction
  • Noncytotoxic and nonhemolytic
  • Sterilized and DNase, RNase, and pyrogen free
  • Packaged in 10 racks of 96
More About this Item
Cole-Parmer sterile pipette tips are constructed of high-grade hydrophobic polypropylene and a polyethylene filter, which prevents liquids and aerosols from entering the body of your pipette. Low retention tips feature a hydrophobic and smooth surface. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
Sterile
Specifications
  • SterileYes
  • Tip Volume (mL)1
  • Body MaterialPolypropylene
  • Wetted MatterialPolypropylene & polyethylene
  • Filter TypePolyethylene
  • DescriptionUniversal Pipette Tips with Filter, Low Retention, Sterile, 1000 μl; 10 Racks x 100 Tips