Cole-Parmer Ultra Flex Support System with 18" Flex Arm and Bench Clamp
Representative image only.
Cole-Parmer Ultra Flex Support System with 18" Flex Arm and Bench Clamp
Cole-Parmer | Item: 0812637
Ultra Flex support system offer the ultimate in flexibility for laboratory use
$581.99CAD / Each
$581.99CAD / Each
Product Description
Key Features
  • Flexible arms are extremely versatile and can be placed in virtually any position or angle
  • Designed to fit easily on benchtops or in fume hoods
  • 2 year warranty
More About this Item
Each Ultra Flex support system includes three interchangeable clamp heads: two-prong clamp head that opens to 2.6" ( 6.6 cm) dia, three-prong clamp head that opens to 2.3" (5.8 cm) dia, spring clamp head that opens to 0.5" (1.3 cm) dia and utility wrench.
Ultra Flex Support System with Bench Clamp
Bench clamp system includes flex arm, interchangeable clamp heads and bench/self clamp. . Designed to securely attach to the side of your lab bench or counter top, to help utilize more space in your lab. Bench clamp made of aluminum
Two year warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Rod MaterialNickel-plated Zinc
  • Base MaterialAluminum
  • Clamp TypeEquipment, Two- or Three-Prong
  • Max Grip Range (InchShort)2.6
  • Length (in)18
  • DescriptionUltra Flex Support System with 18" Flex Arm and Bench Clamp