Cole-Parmer Two-Stage Regulator, 600 scfh Capacity, 350 CGA Fitting
Representative image only.
Cole-Parmer Two-Stage Regulator, 600 scfh Capacity, 350 CGA Fitting
Cole-Parmer | Item: 9820222
$1,193.45CAD / Each
$1,193.45CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Two-stage regulators reduce cylinder pressure in two stages. Use first stage to approach working pressure; use second stage to permit precise adjustment or working pressure. Second stage is overload protected.
1 Year Warranty
Specifications
  • Max Delivery Pressure (psig)50
  • Max Inlet Pressure (PSI)3000
  • Outlet Connections¼" NPT(M)
  • CGA Fitting350
  • Capacity (SCFH)600
  • Outlet Gauge Graduations (PSI)2
  • Inlet Gauge Graduations (PSI)100
  • DescriptionTwo-Stage Regulator, 600 scfh Capacity, 350 CGA Fitting