Cole-Parmer Two-Hole Black Rubber Stoppers, Standard Size 6; 22/Pk
Representative image only.
Cole-Parmer Two-Hole Black Rubber Stoppers, Standard Size 6; 22/Pk
Cole-Parmer | Item: 6299177
$114.92CAD / Pkg of 22
BUY MORE & SAVE
  • 1-5$114.92 CAD / Pkg of 22
  • 6-29$103.43 CAD / Pkg of 22
  • 30-119$97.68 CAD / Pkg of 22
  • 120+$91.94 CAD / Pkg of 22
$114.92CAD / Pkg of 22
BUY MORE & SAVE
  • 1-5$114.92 CAD / Pkg of 22
  • 6-29$103.43 CAD / Pkg of 22
  • 30-119$97.68 CAD / Pkg of 22
  • 120+$91.94 CAD / Pkg of 22
Buy It Together
Current Item
Cole-Parmer Two-Hole Black Rubber Stoppers, Standard Size 6; 22/Pk
Cole-Parmer Two-Hole Black Rubber Stoppers, Standard Size 6; 22/Pk
6299177
$114.92CAD / Pkg of 22
+
Cole-Parmer Twistit black rubber stopper, size 3
6299200
$103.73CAD / Pkg of 50
+ + =
Total Price 372.47 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These two-hole black stoppers made with solid natural rubber stay pliable over a long period of time. Provide good tightness and adhesion, withstand laboratory conditions without hardening, and are ideal for use with nonaggressive or low concentration solutions. Each stopper has its size marked in raised figures. Low sulfur content. Withstand temperatures from –13 to 158°F (–25 to 70°C).
Specifications
  • Standard Size Number6
  • Top Outside Diameter (OD) (mm)32
  • Bottom Outside Diameter (OD) (mm)26
  • Height (mm)25
  • ColorBlack
  • MaterialRubber
  • DescriptionTwo-Hole Black Rubber Stoppers, Standard Size 6; 22/Pk