Cole-Parmer Twistit white rubber stopper assortment pack
Representative image only.
Cole-Parmer Twistit white rubber stopper assortment pack
Cole-Parmer | Item: 6299250
$121.55CAD / Pkg of 33
BUY MORE & SAVE
  • 1-5$121.55 CAD / Pkg of 33
  • 6-29$109.40 CAD / Pkg of 33
  • 30-119$103.32 CAD / Pkg of 33
  • 120+$97.24 CAD / Pkg of 33
$121.55CAD / Pkg of 33
BUY MORE & SAVE
  • 1-5$121.55 CAD / Pkg of 33
  • 6-29$109.40 CAD / Pkg of 33
  • 30-119$103.32 CAD / Pkg of 33
  • 120+$97.24 CAD / Pkg of 33
Buy It Together
Current Item
Cole-Parmer Twistit white rubber stopper assortment pack
Cole-Parmer Twistit white rubber stopper assortment pack
6299250
$121.55CAD / Pkg of 33
+ + =
Total Price 366.72 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Use these versatile Twistit stoppers as either a solid, one-hole, two-hole, or three-hole stopper. Simply twist off a bottom seal to open a hole. The soft resilient natural rubber stays pliable over a long period and withstands laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Available in black or white rubber. Autoclavable.
Assortment pack contains nine size 3,
eight size 4, six size 5, four size 6, two size 7,
two size 8, one size 9, and one size 10
Specifications
  • ColorWhite
  • MaterialRubber
  • DescriptionTwistit white rubber stopper assortment pack