Cole-Parmer Twistit Black Rubber Stopper, Size 9
Representative image only.
Cole-Parmer Twistit Black Rubber Stopper, Size 9
Cole-Parmer | Item: 6299212
$103.73CAD / Pkg of 12
BUY MORE & SAVE
  • 1-5$103.73 CAD / Pkg of 12
  • 6-29$93.36 CAD / Pkg of 12
  • 30-119$88.17 CAD / Pkg of 12
  • 120+$82.98 CAD / Pkg of 12
$103.73CAD / Pkg of 12
BUY MORE & SAVE
  • 1-5$103.73 CAD / Pkg of 12
  • 6-29$93.36 CAD / Pkg of 12
  • 30-119$88.17 CAD / Pkg of 12
  • 120+$82.98 CAD / Pkg of 12
Buy It Together
Current Item
Cole-Parmer Twistit black rubber stopper, size 9
Cole-Parmer Twistit black rubber stopper, size 9
6299212
$103.73CAD / Pkg of 12
+ + =
Total Price 348.90 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Use these versatile Twistit stoppers as either a solid, one-hole, two-hole, or three-hole stopper. Simply twist off a bottom seal to open a hole. The soft resilient natural rubber stays pliable over a long period and withstands laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Available in black or white rubber. Autoclavable.
Specifications
  • Standard Size Number9
  • Top Outside Diameter (OD) (mm)45
  • Bottom Outside Diameter (OD) (mm)37
  • Height (mm)25
  • ColorBlack
  • MaterialRubber
  • DescriptionTwistit black rubber stopper, size 9