Cole-Parmer Threaded to Plug Fitting, Straight Adapter, PVDF, 1/4" NPT(M); 10/PK
Representative image only.
Cole-Parmer Threaded to Plug Fitting, Straight Adapter, PVDF, 1/4" NPT(M); 10/PK
Cole-Parmer | Item: 5014382
High-purity fittings deliver exceptional chemical resistance, durability, and compliance for critical fluid handling in food, bioprocess, and bioresearch applications
$37.87CAD / Pkg of 10
$37.87CAD / Pkg of 10
Product Description
Key Features
  • Injection molded from high-purity, semi-opaque fluoropolymer
  • Provides excellent chemical and corrosion resistance across harsh environments
  • Free from animal-derived materials, plasticizers, phthalates, and latex
  • Stable across a wide temperature range: -35 to 135°C (-31 to 275°F)
  • Supports ultrasonic welding for downstream processing flexibility
  • Sterilize by autoclave, gamma irradiation, or EtO
More About this Item
Engineered from a high-purity thermoplastic fluoropolymer, these PVDF fittings meet the rigorous demands of food, beverage, bioprocess, and bioresearch applications. This advanced material combines outstanding chemical resistance and corrosion protection with exceptional durability, maintaining performance across a wide temperature range from -35 to 135°C (-31 to 275°F).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave, ethylene oxide (EtO), or radiation
  • Chemical ResistanceResistant to most acids, salts, weak bases, halogenated solvents, alcohols, nuclear and UV radiation, oxidants, ozone, and deionized water.
  • Number Of Ports1
  • Connection TypeNPT(M)
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Flow PatternPlug
  • ColorNatural
  • Min Temperature (° F)-31
  • Max Temperature (° F)275
  • Min Temperature (° C)-35
  • Max Temperature (° C)135
  • Regulatory ApprovalsADCF;USP;RoHS
  • Quantity10
  • DescriptionThreaded to Plug Fitting, Straight Adapter, PVDF, 1/4" NPT(M); 10/PK