Cole-Parmer TELOS® WCX 96-well Plate, 30mg / 2mL, 1 / pk

Representative image only.

Cole-Parmer TELOS® WCX 96-well Plate, 30mg / 2mL, 1 / pk

Cole-Parmer | Item: 0647428

Modular design lets you simply remove any unwanted wells and add sealing strips

$530.42CAD / Each

$530.42CAD / Each
Product Description

Key Features

  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Water-wettable polymer is not affected by drying out
  • Applications: Mixed-mode weak cation exchange sorbent for strong basic analyte extraction

More About this Item

Cole-Parmer® TELOS® neo™ populated plates come with 96 wells already in place—ideal for high throughput assays. Polymer-based sorbents have no silanol interactions, excellent flow characteristics, and pH stability providing highly reproducible results. Elution volumes are kept to a minimum by allowing maximum interaction between the analyte and sorbent.

Specifications
  • Bed Mass (mg)30
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsMixed-mode weak cation exchange
  • SorbentWCX
  • DescriptionWCX Fixed-Well Plate, 30 mg sorbent; 1/ea
Reviews
Q&A