Cole-Parmer TELOS® SAX 96-well Plate, 50mg / 2mL, 1 / pk

Representative image only.

Cole-Parmer TELOS® SAX 96-well Plate, 50mg / 2mL, 1 / pk

Cole-Parmer | Item: 0647767

Optimized for sample processing and extraction of small volume biological fluids

$411.08CAD / Pkg of 1

$411.08CAD / Pkg of 1
Product Description

Key Features

  • Select from three main retention mechanisms: nonpolar, ion exchange, or polar
  • Use with vacuum or positive pressure
  • Applications: Ion exchange

More About this Item

The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® Silica-Based SPE fixed 96-well microplates allow for maximum recovery with minimal sample dilution and reduced evaporation time. Select endcapped sorbents for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. C4 (WP) sorbents with average pore size of 300 Å are ideal for compounds with molecular weight >10,000.

Specifications
  • Bed Mass (mg)50
  • Column Particle Size (µm)50
  • End CapNo
  • ApplicationsIon exchange
  • SorbentSAX
Reviews
Q&A