Cole-Parmer Tank Cover, Flat for 2.5 gal Tank 07239-05
Representative image only.
Cole-Parmer Tank Cover, Flat for 2.5 gal Tank 07239-05
Cole-Parmer | Item: 0723960
Reduce evaporation with tank covers
$73.52CAD / Each
BUY MORE & SAVE
  • 1-5$73.52 CAD / Each
  • 6-29$66.17 CAD / Each
  • 30-119$62.49 CAD / Each
  • 120+$58.82 CAD / Each
$73.52CAD / Each
BUY MORE & SAVE
  • 1-5$73.52 CAD / Each
  • 6-29$66.17 CAD / Each
  • 30-119$62.49 CAD / Each
  • 120+$58.82 CAD / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
These stainless steel covers help reduce evaporation, while containing harmful vapors and keeping contaminants out.
Two year warranty
Specifications
  • MaterialStainless steel
  • Compatible WithTank 07239-05
  • DescriptionTank Cover, Flat for 2.5 gal Tank 07239-05