Cole-Parmer Tall-Form Crucible, High Alumina, 250 mL
Representative image only.
Cole-Parmer Tall-Form Crucible, High Alumina, 250 mL
Cole-Parmer | Item: 6310108
Use for high temperature work demanding contamination-free results
$208.85CAD / Each
BUY MORE & SAVE
  • 1-5$208.85 CAD / Each
  • 6-29$187.97 CAD / Each
  • 30-119$177.52 CAD / Each
  • 120+$167.08 CAD / Each
$208.85CAD / Each
BUY MORE & SAVE
  • 1-5$208.85 CAD / Each
  • 6-29$187.97 CAD / Each
  • 30-119$177.52 CAD / Each
  • 120+$167.08 CAD / Each
Buy It Together
Current Item
Cole-Parmer Tall-Form Crucible, High Alumina, 250 mL
Cole-Parmer Tall-Form Crucible, High Alumina, 250 mL
6310108
$208.85CAD / Each
+ + +
Cole-Parmer Lab Jack, 4" x 4" Top, 66 lb Capacity
0805710
$314.57CAD / Each
+ + + + +
Cole-Parmer Crucible Cover, High Alumina, 20 mL
6310162
$14.29CAD / Each
+
Cole-Parmer Crucible Cover, High Alumina, 250 mL
6310168
$24.90CAD / Each
=
Total Price 2045.87 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Highly resistant to chemical attack
  • High alumina ware is highly refractory, ideal for use in reducing and oxidizing atmospheres
  • Autoclavable
  • 1 year warranty
More About this Item
Unglazed tall-form crucibles are 99.7% pure aluminum oxide. Withstand temperatures up to 1750 °C (3182 °F). High alumina labware temperature change rate should not exceed 150 °C/hour.
1 Year Warranty
Specifications
  • MaterialHigh Alumina
  • Capacity (mL)250
  • Top Outside Diameter (mm)88