Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 9; 8/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 9; 8/Pk
Cole-Parmer | Item: 6299130
$180.86CAD / Pkg of 8
BUY MORE & SAVE
  • 1-5$180.86 CAD / Pkg of 8
  • 6-29$162.77 CAD / Pkg of 8
  • 30-119$153.73 CAD / Pkg of 8
  • 120+$144.69 CAD / Pkg of 8
$180.86CAD / Pkg of 8
BUY MORE & SAVE
  • 1-5$180.86 CAD / Pkg of 8
  • 6-29$162.77 CAD / Pkg of 8
  • 30-119$153.73 CAD / Pkg of 8
  • 120+$144.69 CAD / Pkg of 8
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number9
  • Top Outside Diameter (OD) (mm)45
  • Bottom Outside Diameter (OD) (mm)37
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 9; 8/Pk