Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 9.5; 9/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 9.5; 9/Pk
Cole-Parmer | Item: 6299132
$159.13CAD / Pkg of 9
BUY MORE & SAVE
  • 1-5$159.13 CAD / Pkg of 9
  • 6-29$143.22 CAD / Pkg of 9
  • 30-119$135.26 CAD / Pkg of 9
  • 120+$127.30 CAD / Pkg of 9
$159.13CAD / Pkg of 9
BUY MORE & SAVE
  • 1-5$159.13 CAD / Pkg of 9
  • 6-29$143.22 CAD / Pkg of 9
  • 30-119$135.26 CAD / Pkg of 9
  • 120+$127.30 CAD / Pkg of 9
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number9.5
  • Top Outside Diameter (OD) (mm)46
  • Bottom Outside Diameter (OD) (mm)38
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 9.5; 9/Pk