Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 8.5; 11/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 8.5; 11/Pk
Cole-Parmer | Item: 6299128
$164.28CAD / Pkg of 11
BUY MORE & SAVE
  • 1-5$164.28 CAD / Pkg of 11
  • 6-29$147.85 CAD / Pkg of 11
  • 30-119$139.64 CAD / Pkg of 11
  • 120+$131.42 CAD / Pkg of 11
$164.28CAD / Pkg of 11
BUY MORE & SAVE
  • 1-5$164.28 CAD / Pkg of 11
  • 6-29$147.85 CAD / Pkg of 11
  • 30-119$139.64 CAD / Pkg of 11
  • 120+$131.42 CAD / Pkg of 11
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number8.5
  • Top Outside Diameter (OD) (mm)43
  • Bottom Outside Diameter (OD) (mm)36
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 8.5; 11/Pk