Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 7; 12/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 7; 12/Pk
Cole-Parmer | Item: 6299122
$129.95CAD / Pkg of 12
BUY MORE & SAVE
  • 1-5$129.95 CAD / Pkg of 12
  • 6-29$116.96 CAD / Pkg of 12
  • 30-119$110.46 CAD / Pkg of 12
  • 120+$103.96 CAD / Pkg of 12
$129.95CAD / Pkg of 12
BUY MORE & SAVE
  • 1-5$129.95 CAD / Pkg of 12
  • 6-29$116.96 CAD / Pkg of 12
  • 30-119$110.46 CAD / Pkg of 12
  • 120+$103.96 CAD / Pkg of 12
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number7
  • Top Outside Diameter (OD) (mm)37
  • Bottom Outside Diameter (OD) (mm)30
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 7; 12/Pk