Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 6.5; 15/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 6.5; 15/Pk
Cole-Parmer | Item: 6299120
$160.60CAD / Pkg of 15
BUY MORE & SAVE
  • 1-5$160.60 CAD / Pkg of 15
  • 6-29$144.54 CAD / Pkg of 15
  • 30-119$136.51 CAD / Pkg of 15
  • 120+$128.48 CAD / Pkg of 15
$160.60CAD / Pkg of 15
BUY MORE & SAVE
  • 1-5$160.60 CAD / Pkg of 15
  • 6-29$144.54 CAD / Pkg of 15
  • 30-119$136.51 CAD / Pkg of 15
  • 120+$128.48 CAD / Pkg of 15
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number6.5
  • Top Outside Diameter (OD) (mm)34
  • Bottom Outside Diameter (OD) (mm)27
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 6.5; 15/Pk