Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 6; 17/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 6; 17/Pk
Cole-Parmer | Item: 6299118
$141.30CAD / Pkg of 17
BUY MORE & SAVE
  • 1-5$141.30 CAD / Pkg of 17
  • 6-29$127.17 CAD / Pkg of 17
  • 30-119$120.11 CAD / Pkg of 17
  • 120+$113.04 CAD / Pkg of 17
$141.30CAD / Pkg of 17
BUY MORE & SAVE
  • 1-5$141.30 CAD / Pkg of 17
  • 6-29$127.17 CAD / Pkg of 17
  • 30-119$120.11 CAD / Pkg of 17
  • 120+$113.04 CAD / Pkg of 17
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number6
  • Top Outside Diameter (OD) (mm)32
  • Bottom Outside Diameter (OD) (mm)26
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 6; 17/Pk