Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 5.5; 23/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 5.5; 23/Pk
Cole-Parmer | Item: 6299116
$156.18CAD / Pkg of 23
BUY MORE & SAVE
  • 1-5$156.18 CAD / Pkg of 23
  • 6-29$140.56 CAD / Pkg of 23
  • 30-119$132.75 CAD / Pkg of 23
  • 120+$124.94 CAD / Pkg of 23
$156.18CAD / Pkg of 23
BUY MORE & SAVE
  • 1-5$156.18 CAD / Pkg of 23
  • 6-29$140.56 CAD / Pkg of 23
  • 30-119$132.75 CAD / Pkg of 23
  • 120+$124.94 CAD / Pkg of 23
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number5.5
  • Top Outside Diameter (OD) (mm)28
  • Bottom Outside Diameter (OD) (mm)24
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 5.5; 23/Pk