Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 4; 27/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 4; 27/Pk
Cole-Parmer | Item: 6299112
$129.95CAD / Pkg of 27
BUY MORE & SAVE
  • 1-5$129.95 CAD / Pkg of 27
  • 6-29$116.96 CAD / Pkg of 27
  • 30-119$110.46 CAD / Pkg of 27
  • 120+$103.96 CAD / Pkg of 27
$129.95CAD / Pkg of 27
BUY MORE & SAVE
  • 1-5$129.95 CAD / Pkg of 27
  • 6-29$116.96 CAD / Pkg of 27
  • 30-119$110.46 CAD / Pkg of 27
  • 120+$103.96 CAD / Pkg of 27
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number4
  • Top Outside Diameter (OD) (mm)26
  • Bottom Outside Diameter (OD) (mm)20
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 4; 27/Pk