Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 3; 32/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 3; 32/Pk
Cole-Parmer | Item: 6299110
$131.72CAD / Pkg of 32
BUY MORE & SAVE
  • 1-5$131.72 CAD / Pkg of 32
  • 6-29$118.55 CAD / Pkg of 32
  • 30-119$111.96 CAD / Pkg of 32
  • 120+$105.38 CAD / Pkg of 32
$131.72CAD / Pkg of 32
BUY MORE & SAVE
  • 1-5$131.72 CAD / Pkg of 32
  • 6-29$118.55 CAD / Pkg of 32
  • 30-119$111.96 CAD / Pkg of 32
  • 120+$105.38 CAD / Pkg of 32
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number3
  • Top Outside Diameter (OD) (mm)24
  • Bottom Outside Diameter (OD) (mm)18
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 3; 32/Pk