Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 2; 44/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 2; 44/Pk
Cole-Parmer | Item: 6299108
$142.62CAD / Pkg of 44
BUY MORE & SAVE
  • 1-5$142.62 CAD / Pkg of 44
  • 6-29$128.36 CAD / Pkg of 44
  • 30-119$121.23 CAD / Pkg of 44
  • 120+$114.10 CAD / Pkg of 44
$142.62CAD / Pkg of 44
BUY MORE & SAVE
  • 1-5$142.62 CAD / Pkg of 44
  • 6-29$128.36 CAD / Pkg of 44
  • 30-119$121.23 CAD / Pkg of 44
  • 120+$114.10 CAD / Pkg of 44
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number2
  • Top Outside Diameter (OD) (mm)20
  • Bottom Outside Diameter (OD) (mm)16
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 2; 44/Pk