Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 12; 5/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 12; 5/Pk
Cole-Parmer | Item: 6299142
$172.39CAD / Pkg of 5
BUY MORE & SAVE
  • 1-5$172.39 CAD / Pkg of 5
  • 6-29$155.15 CAD / Pkg of 5
  • 30-119$146.53 CAD / Pkg of 5
  • 120+$137.91 CAD / Pkg of 5
$172.39CAD / Pkg of 5
BUY MORE & SAVE
  • 1-5$172.39 CAD / Pkg of 5
  • 6-29$155.15 CAD / Pkg of 5
  • 30-119$146.53 CAD / Pkg of 5
  • 120+$137.91 CAD / Pkg of 5
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number12
  • Top Outside Diameter (OD) (mm)64
  • Bottom Outside Diameter (OD) (mm)54
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 12; 5/Pk