Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 11; 6/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 11; 6/Pk
Cole-Parmer | Item: 6299138
$134.08CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$134.08 CAD / Pkg of 6
  • 6-29$120.67 CAD / Pkg of 6
  • 30-119$113.97 CAD / Pkg of 6
  • 120+$107.26 CAD / Pkg of 6
$134.08CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$134.08 CAD / Pkg of 6
  • 6-29$120.67 CAD / Pkg of 6
  • 30-119$113.97 CAD / Pkg of 6
  • 120+$107.26 CAD / Pkg of 6
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number11
  • Top Outside Diameter (OD) (mm)56
  • Bottom Outside Diameter (OD) (mm)48
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 11; 6/Pk