Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 10; 8/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 10; 8/Pk
Cole-Parmer | Item: 6299134
$137.03CAD / Pkg of 8
BUY MORE & SAVE
  • 1-5$137.03 CAD / Pkg of 8
  • 6-29$123.33 CAD / Pkg of 8
  • 30-119$116.48 CAD / Pkg of 8
  • 120+$109.62 CAD / Pkg of 8
$137.03CAD / Pkg of 8
BUY MORE & SAVE
  • 1-5$137.03 CAD / Pkg of 8
  • 6-29$123.33 CAD / Pkg of 8
  • 30-119$116.48 CAD / Pkg of 8
  • 120+$109.62 CAD / Pkg of 8
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number10
  • Top Outside Diameter (OD) (mm)50
  • Bottom Outside Diameter (OD) (mm)42
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 10; 8/Pk