Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 10.5; 7/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 10.5; 7/Pk
Cole-Parmer | Item: 6299136
$162.07CAD / Pkg of 7
BUY MORE & SAVE
  • 1-5$162.07 CAD / Pkg of 7
  • 6-29$145.86 CAD / Pkg of 7
  • 30-119$137.76 CAD / Pkg of 7
  • 120+$129.66 CAD / Pkg of 7
$162.07CAD / Pkg of 7
BUY MORE & SAVE
  • 1-5$162.07 CAD / Pkg of 7
  • 6-29$145.86 CAD / Pkg of 7
  • 30-119$137.76 CAD / Pkg of 7
  • 120+$129.66 CAD / Pkg of 7
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number10.5
  • Top Outside Diameter (OD) (mm)53
  • Bottom Outside Diameter (OD) (mm)45
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 10.5; 7/Pk