Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 1; 48/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 1; 48/Pk
Cole-Parmer | Item: 6299106
$121.26CAD / Pkg of 48
BUY MORE & SAVE
  • 1-5$121.26 CAD / Pkg of 48
  • 6-29$109.13 CAD / Pkg of 48
  • 30-119$103.07 CAD / Pkg of 48
  • 120+$97.01 CAD / Pkg of 48
$121.26CAD / Pkg of 48
BUY MORE & SAVE
  • 1-5$121.26 CAD / Pkg of 48
  • 6-29$109.13 CAD / Pkg of 48
  • 30-119$103.07 CAD / Pkg of 48
  • 120+$97.01 CAD / Pkg of 48
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number1
  • Top Outside Diameter (OD) (mm)19
  • Bottom Outside Diameter (OD) (mm)14
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 1; 48/Pk