Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 00; 95/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 00; 95/Pk
Cole-Parmer | Item: 6299102
$153.60CAD / Pkg of 95
BUY MORE & SAVE
  • 1-5$153.60 CAD / Pkg of 95
  • 6-29$138.24 CAD / Pkg of 95
  • 30-119$130.56 CAD / Pkg of 95
  • 120+$122.88 CAD / Pkg of 95
$153.60CAD / Pkg of 95
BUY MORE & SAVE
  • 1-5$153.60 CAD / Pkg of 95
  • 6-29$138.24 CAD / Pkg of 95
  • 30-119$130.56 CAD / Pkg of 95
  • 120+$122.88 CAD / Pkg of 95
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number00
  • Top Outside Diameter (OD) (mm)15
  • Bottom Outside Diameter (OD) (mm)10
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 00; 95/Pk