Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 9.5, Pink; 5/Bag
Representative image only.
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 9.5, Pink; 5/Bag
Cole-Parmer | Item: 0629816
$131.72CAD / Bag of 5
BUY MORE & SAVE
  • 1-5$131.72 CAD / Bag of 5
  • 6-29$118.55 CAD / Bag of 5
  • 30-119$111.96 CAD / Bag of 5
  • 120+$105.38 CAD / Bag of 5
$131.72CAD / Bag of 5
BUY MORE & SAVE
  • 1-5$131.72 CAD / Bag of 5
  • 6-29$118.55 CAD / Bag of 5
  • 30-119$111.96 CAD / Bag of 5
  • 120+$105.38 CAD / Bag of 5
Product Description
Key Features
  • 6 month warranty
More About this Item
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers.
Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*
6 Month Warranty
Specifications
  • Standard Size Number9.5
  • Top Outside Diameter (OD) (mm)46
  • Bottom Outside Diameter (OD) (mm)38
  • Height (mm)42
  • ColorPink
  • MaterialSilicone
  • DescriptionSolid Color-Coded Silicone Stoppers, Standard Size 9.5, Pink; 5/Bag