Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 8, Green; 10/Bag
Representative image only.
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 8, Green; 10/Bag
Cole-Parmer | Item: 0629814
$185.65CAD / Bag of 10
BUY MORE & SAVE
  • 1-5$185.65 CAD / Bag of 10
  • 6-29$167.09 CAD / Bag of 10
  • 30-119$157.80 CAD / Bag of 10
  • 120+$148.52 CAD / Bag of 10
$185.65CAD / Bag of 10
BUY MORE & SAVE
  • 1-5$185.65 CAD / Bag of 10
  • 6-29$167.09 CAD / Bag of 10
  • 30-119$157.80 CAD / Bag of 10
  • 120+$148.52 CAD / Bag of 10
Product Description
Key Features
  • 6 month warranty
More About this Item
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers.
Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*
6 Month Warranty
Specifications
  • Standard Size Number8
  • Top Outside Diameter (OD) (mm)40
  • Bottom Outside Diameter (OD) (mm)33
  • Height (mm)40
  • ColorGreen
  • MaterialSilicone
  • DescriptionSolid Color-Coded Silicone Stoppers, Standard Size 8, Green; 10/Bag