Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 11, White; 5/Bag
Representative image only.
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 11, White; 5/Bag
Cole-Parmer | Item: 0629820
$250.11CAD / Bag of 5
BUY MORE & SAVE
  • 1-5$250.11 CAD / Bag of 5
  • 6-29$225.10 CAD / Bag of 5
  • 30-119$212.59 CAD / Bag of 5
  • 120+$200.09 CAD / Bag of 5
$250.11CAD / Bag of 5
BUY MORE & SAVE
  • 1-5$250.11 CAD / Bag of 5
  • 6-29$225.10 CAD / Bag of 5
  • 30-119$212.59 CAD / Bag of 5
  • 120+$200.09 CAD / Bag of 5
Product Description
Key Features
  • 6 month warranty
More About this Item
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers.
Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*
6 Month Warranty
Specifications
  • Standard Size Number11
  • Top Outside Diameter (OD) (mm)58
  • Bottom Outside Diameter (OD) (mm)49
  • Height (mm)52
  • ColorWhite
  • MaterialSilicone
  • DescriptionSolid Color-Coded Silicone Stoppers, Standard Size 11, White; 5/Bag