Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 10.5, Blue; 5/Bag
Representative image only.
Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 10.5, Blue; 5/Bag
Cole-Parmer | Item: 0629818
$215.48CAD / Bag of 5
BUY MORE & SAVE
  • 1-5$215.48 CAD / Bag of 5
  • 6-29$193.93 CAD / Bag of 5
  • 30-119$183.16 CAD / Bag of 5
  • 120+$172.38 CAD / Bag of 5
$215.48CAD / Bag of 5
BUY MORE & SAVE
  • 1-5$215.48 CAD / Bag of 5
  • 6-29$193.93 CAD / Bag of 5
  • 30-119$183.16 CAD / Bag of 5
  • 120+$172.38 CAD / Bag of 5
Product Description
Key Features
  • 6 month warranty
More About this Item
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers.
Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*
6 Month Warranty
Specifications
  • Standard Size Number10.5
  • Top Outside Diameter (OD) (mm)52
  • Bottom Outside Diameter (OD) (mm)44
  • Height (mm)48
  • ColorBlue
  • MaterialSilicone
  • DescriptionSolid Color-Coded Silicone Stoppers, Standard Size 10.5, Blue; 5/Bag