Cole-Parmer Sit-Stand Stool, Polyurethane, Bench Height, Black, Chrome Casters, Chrome Foot Ring
Representative image only.
Cole-Parmer Sit-Stand Stool, Polyurethane, Bench Height, Black, Chrome Casters, Chrome Foot Ring
Cole-Parmer | Item: 9000541
Sit comfortably, stand quickly and adjust the height easily
$461.91CAD / Each
BUY MORE & SAVE
  • 1-3$461.91 CAD / Each
  • 4-9$415.72 CAD / Each
  • 10+$392.62 CAD / Each
$461.91CAD / Each
BUY MORE & SAVE
  • 1-3$461.91 CAD / Each
  • 4-9$415.72 CAD / Each
  • 10+$392.62 CAD / Each
Product Description
Key Features
  • Rugged polyurethane tractor seat is chemical- and puncture-resistant
  • Constructed with 26-in. 5-leg polished cast aluminum base for enhanced durability
  • Rugged and durable weight capacity of 300 lbs.
  • Bench height chairs feature an 21 to 29" pneumatic adjustment
  • Chrome hooded, rolling, non-marring casters are suitable for both hard and carpeted floors
  • Constructed with 20” (50.8 cm) diameter chrome foot ring
  • Warranty 15-years parts, lifetime pneumatic
More About this Item
These ergonomic polyurethane tractor stools are constructed from our rugged polyurethane that is self-skinning, chemical- and puncture-resistant, and fully washable.
All stools feature a smooth pneumatic lift. Choose from desk or bench height chairs with pneumatic adjustment. Chairs are also available in black or blue colors and with or without foot ring. Choose casters or glides, both of which are non-marring and suitable for hard or carpeted floors.
Specifications
  • Dimension17-22" adjustable height (measured from the top of the seat to the floor)
  • MaterialPolyurethane
  • ColorBlack
  • DescriptionSit-Stand Stool, Polyurethane, Bench Height, Black, Chrome Casters, Chrome Foot Ring
  • WarrantyWarranty 15-years parts, 5-years upholstery, lifetime pneumatic