Cole-Parmer Silicone Bath Fluid; 5 Gallons

Representative image only.
Cole-Parmer Item # RK-78930-20
Ideal for high oxidation applications
  • Strong resistance to oxidation up to 204°C (400°F)
  • Maximum operating temperature of 316°C (600°F)
  • Pumpable down to -50°C (-46°F)
  • ASTM D92 flash point at 324°C

$1,508.75CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° C)204
  • DescriptionSilicone Bath Fluid - 5 Gallon Pail

Key Features

  • Strong resistance to oxidation up to 204°C (400°F)
  • Maximum operating temperature of 316°C (600°F)
  • Pumpable down to -50°C (-46°F)
  • ASTM D92 flash point at 324°C

More About this Item

This silicon-based transfer bath fluid is best suited for high oxidation applications and can be use in fully exposed systems like open baths. Resistant to fouling, you can expect to get a long service life from the fuel. It is ideal for applications not prone to contamination or leaking. The non-corrosive and non-toxic thermal fluid is safe to use.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat