Cole-Parmer SamplePrep MiniG® Homogenizer Foam Block, 12 x 7 mL Centrifuge Tubes; 1/PK
Representative image only.
Cole-Parmer SamplePrep MiniG® Homogenizer Foam Block, 12 x 7 mL Centrifuge Tubes; 1/PK
Cole-Parmer SamplePrep | Mfr: 1683| Item: 0557538
Accommodates a full range of sample vials with an adjustable clamp
$218.06CAD / Pkg of 1
$218.06CAD / Pkg of 1
Buy It Together
Current Item
Cole-Parmer SamplePrep MiniG® Homogenizer Foam Block, 12 x 7 mL Centrifuge Tubes; 1/PK
Cole-Parmer SamplePrep MiniG® Homogenizer Foam Block, 12 x 7 mL Centrifuge Tubes; 1/PK
0557538
$218.06CAD / Pkg of 1
+ + =
Total Price 331.51 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Rugged foam block holds 12, 7 mL centrifuge tubes
  • One pair per package
More About this Item
Multiple inserts are available for a variety of homogenizing applications.
Specifications
  • Use with GeneratorsMiniG® Homogenizer
  • MaterialFoam
  • DescriptionMiniG® Homogenizer Foam Block, 12 x 7 mL Centrifuge Tubes; 1/PK