Cole-Parmer Reversible 16-Point Vertical Temperature Label Black/Red, 32-49C/90-120F; 10/Pk
Representative image only.
Cole-Parmer Reversible 16-Point Vertical Temperature Label Black/Red, 32-49C/90-120F; 10/Pk
Cole-Parmer | Item: 0903555
Each style covers a wide range of temperatures, for use in many different application
$57.31CAD / Pkg of 10
BUY MORE & SAVE
  • 1-4$57.31 CAD / Pkg of 10
  • 5-9$51.58 CAD / Pkg of 10
  • 10-19$48.71 CAD / Pkg of 10
  • 20+$45.85 CAD / Pkg of 10
$57.31CAD / Pkg of 10
BUY MORE & SAVE
  • 1-4$57.31 CAD / Pkg of 10
  • 5-9$51.58 CAD / Pkg of 10
  • 10-19$48.71 CAD / Pkg of 10
  • 20+$45.85 CAD / Pkg of 10
Product Description
Key Features
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.
1 Year Warranty
Specifications
  • Number of Temperature Indicators16
  • Max temperature (° F)120
  • Max temperature (° C)49
  • Temperature Points (° F)90, 92, 94, 96, 98, 100, 102, 104, 106, 108, 110, 112, 114, 116, 118, 120
  • Temperature Points (° C)32, 33, 34, 35, 37, 38, 39, 40, 41, 42, 43, 44, 45, 47, 48, 49
  • Initial colorBlack
  • First color changeRed
  • Length (in)1/2
  • Depth (in)5
  • DescriptionReversible 16-Point Vertical Temperature Label Black/Red, 32-49C/90-120F; 10/Pk