Cole-Parmer Replacement Carboy Spigot/Stopcock
Representative image only.
Cole-Parmer Replacement Carboy Spigot/Stopcock
Cole-Parmer | Item: 6250790
Convenient replacement part for spent or damaged spigot
$59.38CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$59.38 CAD / Pkg of 1
  • 6-29$53.44 CAD / Pkg of 1
  • 30-119$50.47 CAD / Pkg of 1
  • 120+$47.50 CAD / Pkg of 1
$59.38CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$59.38 CAD / Pkg of 1
  • 6-29$53.44 CAD / Pkg of 1
  • 30-119$50.47 CAD / Pkg of 1
  • 120+$47.50 CAD / Pkg of 1
Buy It Together
Current Item
Cole-Parmer Replacement Carboy Spigot/Stopcock
Cole-Parmer Replacement Carboy Spigot/Stopcock
6250790
$59.38CAD / Pkg of 1
+
Cole-Parmer Economical Heavy Duty Carboy with Spigot, 5 L
6250705
$152.86CAD / Pkg of 1
=
Total Price 212.24 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Removeable tube fitting accepts 3/8" (8 mm) inner diameter flexible tubing
  • Silicone o-ring
  • Autoclavable
  • 1 year warranty
More About this Item
Replacement polypropylene spigot with silicone o-ring for the Cole-Parmer Essentials economical heavy-duty carboys.
Please note: The recommended autoclave cycle for empty containers is 121 °C standard setting for 15 minutes. Care must be taken to allow free air circulation into and out of vessels during the autoclave cycle, especially during the venting and cooling stages.
1 Year Warranty
Specifications
  • MaterialPP/Silicone
  • Compatible With62507-05, 62507-15, or 62507-25
  • Diameter (InchShort)3/8
  • DescriptionReplacement Carboy Spigot/Stopcock; 1/Pk