Cole-Parmer PTFE Stirrer Bars, Boxed Assortments, 18 Plain, 2 each of 10 x 6, 15 x 6, 20 x 7, 25 x 8, 30 x 7, 40 x 8, 50 x 8, 60 x 7, 80 x 10 mm
Representative image only.
Cole-Parmer PTFE Stirrer Bars, Boxed Assortments, 18 Plain, 2 each of 10 x 6, 15 x 6, 20 x 7, 25 x 8, 30 x 7, 40 x 8, 50 x 8, 60 x 7, 80 x 10 mm
Cole-Parmer | Item: 7890341
Boxed sets include an assortment of useful sizes in a re-usable container
$296.52CAD / Each
BUY MORE & SAVE
  • 1-9$296.52 CAD / Each
  • 10-24$266.87 CAD / Each
  • 25-49$252.04 CAD / Each
  • 50+$237.22 CAD / Each
$296.52CAD / Each
BUY MORE & SAVE
  • 1-9$296.52 CAD / Each
  • 10-24$266.87 CAD / Each
  • 25-49$252.04 CAD / Each
  • 50+$237.22 CAD / Each
Product Description
Key Features
  • Isostatic encapsulation eliminates cracks and porosity
  • FDA and USP Class VI approved PTFE
  • Polished finish reduces pick-up and cross contamination
More About this Item
Magnetic stirring is a widely used method for stirring and mixing in liquid media. The process is not only simple and inexpensive, but extremely diverse in the range of application.
Specifications
  • DescriptionPTFE Stirrer Bars, Boxed Assortments, 18 Plain, 2 each of 10 x 6, 15 x 6, 20 x 7, 25 x 8, 30 x 7, 40 x 8, 50 x 8, 60 x 7, 80 x 10 mm