Cole-Parmer PTFE Beaker Cover for 06300-96
Representative image only.
Cole-Parmer PTFE Beaker Cover for 06300-96
Cole-Parmer | Item: 0630099
Stand up to the harshest chemicals
$264.47CAD / Each
BUY MORE & SAVE
  • 1-5$264.47 CAD / Each
  • 6-29$238.02 CAD / Each
  • 30-119$224.80 CAD / Each
  • 120+$211.58 CAD / Each
$264.47CAD / Each
BUY MORE & SAVE
  • 1-5$264.47 CAD / Each
  • 6-29$238.02 CAD / Each
  • 30-119$224.80 CAD / Each
  • 120+$211.58 CAD / Each
Product Description
Key Features
  • Constructed of chemically inert PTFE
  • Autoclavable
  • Beaker cover is suitable for 5000 mL beaker 06300-96
More About this Item
These PTFE beaker covers are compatible with the 06300-series PTFE broad spout beakers.
Specifications
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap MaterialPolytetrafluoroethylene (PTFE)
  • Cap ColorWhite
  • Withstands Low Temperature (° F)-328
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)572
  • Outside Diameter (OD) (cm)20
  • DescriptionPTFE Beaker Cover for 06300-96, 1/ea