Cole-Parmer PTFE Beaker Cover for 06300-62; 2/PK
Representative image only.
Cole-Parmer PTFE Beaker Cover for 06300-62; 2/PK
Cole-Parmer | Item: 0630079
Stand up to the harshest chemicals
$35.36CAD / Pkg of 2
BUY MORE & SAVE
  • 1-5$35.36 CAD / Pkg of 2
  • 6-29$31.82 CAD / Pkg of 2
  • 30-119$30.06 CAD / Pkg of 2
  • 120+$28.29 CAD / Pkg of 2
$35.36CAD / Pkg of 2
BUY MORE & SAVE
  • 1-5$35.36 CAD / Pkg of 2
  • 6-29$31.82 CAD / Pkg of 2
  • 30-119$30.06 CAD / Pkg of 2
  • 120+$28.29 CAD / Pkg of 2
Product Description
Key Features
  • Constructed of chemically inert PTFE
  • Autoclavable
  • Beaker cover is suitable for 1 mL beaker 06300-62
More About this Item
These PTFE beaker covers are compatible with the 06300-series PTFE broad spout beakers.
Specifications
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap MaterialPolytetrafluoroethylene (PTFE)
  • Cap ColorWhite
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)572
  • Withstands Low Temperature (° F)-328
  • Outside Diameter (OD) (cm)2
  • DescriptionPTFE Beaker Cover for 06300-62, 2/ea