Cole-Parmer pHPerch benchtop workstation; small (White)

Representative image only.

Cole-Parmer pHPerch benchtop workstation; small (White)

Cole-Parmer | Item: 6352730

$438.33CAD / Each

$438.33CAD / Each
Product Description

Key Features

  • Keep buffers, wash bottles, and waste beakers organized and in easy reach.
  • 1 year warranty
1 Year Warranty
Specifications
  • Number Of Compartments10
  • ColorWhite
  • Exterior Width (in)13
  • Exterior Height (in)12
  • Exterior Depth (in)6
  • Exterior Width (cm)33.02
  • Exterior Height (cm)30.48
  • Exterior Depth (cm)15.24
  • DescriptionpHPerch benchtop workstation; small (White)
  • Warranty1 year
Reviews
Q&A