Cole-Parmer Nipple Fittings, Straight Union, PP, 1/8" NPT(M); 10/PK
Representative image only.
Cole-Parmer Nipple Fittings, Straight Union, PP, 1/8" NPT(M); 10/PK
Cole-Parmer | Item: 5014402
High-purity fittings deliver exceptional chemical resistance, durability, and compliance for critical fluid handling in food, bioprocess, and bioresearch applications
$11.93CAD / Pkg of 10
$11.93CAD / Pkg of 10
Product Description
Key Features
  • Injection molded from high-purity, semi-opaque fluoropolymer
  • Provides excellent chemical and corrosion resistance across harsh environments
  • Free from animal-derived materials, plasticizers, phthalates, and latex
  • Stable across a wide temperature range: -35 to 135°C (-31 to 275°F)
  • Supports ultrasonic welding for downstream processing flexibility
  • Sterilize by autoclave, gamma irradiation, or EtO
More About this Item
Engineered from a high-purity thermoplastic fluoropolymer, these PVDF fittings meet the rigorous demands of food, beverage, bioprocess, and bioresearch applications. This advanced material combines outstanding chemical resistance and corrosion protection with exceptional durability, maintaining performance across a wide temperature range from -35 to 135°C (-31 to 275°F).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Chemical ResistanceResistant to solvents, chemicals, water, and other inorganic environments; resists most strong mineral acids and bases; subject to attack by oxidizing agents.
  • Connection TypeNPT(M) x NPT(M)
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Thread Size NPT(M) (Port 2) (InchShort)1/8
  • Flow PatternStraight
  • Regulatory ApprovalsFDA;ADCF;RoHS
  • Quantity10
  • DescriptionCole-Parmer Nipple Fittings, 1/8" NPTM PP, 10PK