Cole-Parmer Microcentrifuge Tube with Cap, Non-Sterile, Polypropylene, Conical Bottom, 1.5 mL; 500/PK

Representative image only.

Cole-Parmer Microcentrifuge Tube with Cap, Non-Sterile, Polypropylene, Conical Bottom, 1.5 mL; 500/PK

Cole-Parmer | Item: 0634462

Easy to open microcentrifuge tubes with backspring flip cap eliminates sore thumbs

$335.20CAD / Pkg of 500

$335.20CAD / Pkg of 500
Product Description

Key Features

  • Small tab at tube back for easy, comfortable cap opening
  • Safely centrifuge up to 25,000 x g
  • Excellent sealing performance
  • DNase/RNase free, nonpyrogenic
  • Autoclavable

More About this Item

Reduce the stress on your hand due to repeatedly opening and closing microcentrifuge tubes. This unique 1.5 mL tube opens by pushing a tab at the back of the cap allowing for easy, one-handed operation. Useful for processing and storage of small volume samples, reagents and extracts, they have a MAX RCF (maximum relative centrifugal force) of 25,000 x g. The smooth, transparent tube body has clear graduations from 0.1 to 1.5 mL in 0.25 mL increments, and a frosted marking area for easy tagging and identification.

These conical bottom microcentrifuge tubes are made of high-quality polymer polypropylene (PP), are strictly tested for leakage, and have excellent sealing performance. They withstand temperatures from -80 °C (-112 °F) to 121 °C (250 °F), and are DNase/RNase free, nonpyrogenic, and conform to USP Class VI standards. Packaged in resealable bags of 500, they have a printed lot number for quality traceability.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

BioPyr
Specifications
  • Max RCF x g25000
  • SterileNo
  • AutoclavableYes
  • Tube Capacity (mL)1.5
  • Tube MaterialPolypropylene (PP)
  • Tube ColorClear
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleBackSpring
  • Cap MaterialPolypropylene (PP)
  • Graduations0.1 to 1.5 in 0.25 mL increments
  • Packaging500/PK
  • DescriptionMicrocentrifuge Tube with BackSpring Cap, Non-Sterile, Polypropylene, Conical Bottom, 1.5 mL; 500/PK
Reviews
Q&A