Cole-Parmer KIT 1 UV/Vis Photometric Accuracy and Stray Light Calibration Kit
Representative image only.
Cole-Parmer KIT 1 UV/Vis Photometric Accuracy and Stray Light Calibration Kit
Cole-Parmer | Item: 0266001
Easily and accurately perform certified spectrophotometer evaluations in any visible or UV-visible spectrophotometer!
$8,464.63CAD / Each
$8,464.63CAD / Each
Product Description
Includes
one hard plastic carrying case.
Key Features
  • Standards are durable, scratch-resistant and never need to be replaced
  • ISO 17025 accredited for testing and calibration and NIST compatible
  • 1 year warranty
More About this Item
Verify the performance and accuracy of your spectrophotometer right in your own lab. These permanent standards do not have to be recalibrated due to material aging. Unique material construction is scratch resistant and provides stable optical performance.
Photometric Accuracy and Stray Light Calibration Kit 02660-01—use for UV-VIS photometric accuracy, absorbance accuracy, and stray light between 400 to 700 nm.
1 Year Warranty
Specifications
  • Stray LightYes
  • Photometric AccuracyYes
  • DescriptionUV/Vis Photometric Accuracy and Stray Light Calibration Kit