Cole-Parmer Irreversible 5 to Point Vertical Temperature Label, 171 to 210°F/77 to 99°C; 10/Pk
Representative image only.
Cole-Parmer Irreversible 5 to Point Vertical Temperature Label, 171 to 210°F/77 to 99°C; 10/Pk
Cole-Parmer | Item: 0903519
Reliable proof that the product was maintained at the correct temperature
$55.40CAD / Pkg of 10
BUY MORE & SAVE
  • 1-4$55.40 CAD / Pkg of 10
  • 5-9$49.86 CAD / Pkg of 10
  • 10-19$47.09 CAD / Pkg of 10
  • 20+$44.32 CAD / Pkg of 10
$55.40CAD / Pkg of 10
BUY MORE & SAVE
  • 1-4$55.40 CAD / Pkg of 10
  • 5-9$49.86 CAD / Pkg of 10
  • 10-19$47.09 CAD / Pkg of 10
  • 20+$44.32 CAD / Pkg of 10
Product Description
Key Features
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
  • 1 year warranty
More About this Item
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F.
1 Year Warranty
Specifications
  • Number of Temperature Indicators5
  • Max temperature (° F)210
  • Max temperature (° C)99
  • Temperature Points (° F)171, 180, 180, 199, 210
  • Temperature Points (° C)77, 82, 88, 93, 99
  • Initial colorWhite
  • First color changeBlack
  • Width (in)45/64
  • Length (in)1 17/32
  • Depth (in)1/32
  • DescriptionIrreversible 5 to Point Vertical Temperature Label, 171 to 210°F/77 to 99°C; 10/Pk