Cole-Parmer High-purity silicone stopper, size 7 (38D)
Representative image only.
Cole-Parmer High-purity silicone stopper, size 7 (38D)
Cole-Parmer | Item: 6299426
Stop particle buildup
$98.57CAD / Pkg of 5
BUY MORE & SAVE
  • 1-5$98.57 CAD / Pkg of 5
  • 6-29$88.71 CAD / Pkg of 5
  • 30-119$83.78 CAD / Pkg of 5
  • 120+$78.86 CAD / Pkg of 5
$98.57CAD / Pkg of 5
BUY MORE & SAVE
  • 1-5$98.57 CAD / Pkg of 5
  • 6-29$88.71 CAD / Pkg of 5
  • 30-119$83.78 CAD / Pkg of 5
  • 120+$78.86 CAD / Pkg of 5
Product Description
Key Features
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
More About this Item
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. Autoclavable.
6 Month Warranty
Specifications
  • Standard Size Number7
  • European Size Number38D
  • Top Outside Diameter (OD) (mm)38
  • Bottom Outside Diameter (OD) (mm)31
  • Height (mm)35
  • ColorTranslucent
  • MaterialSilicone
  • DescriptionHigh-purity silicone stopper, size 7 (38D)