Cole-Parmer High-purity silicone stopper, size 13 (70D)
Representative image only.
Cole-Parmer High-purity silicone stopper, size 13 (70D)
Cole-Parmer | Item: 6299440
Stop particle buildup
$110.95CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$110.95 CAD / Pkg of 1
  • 6-29$99.86 CAD / Pkg of 1
  • 30-119$94.31 CAD / Pkg of 1
  • 120+$88.76 CAD / Pkg of 1
$110.95CAD / Pkg of 1
BUY MORE & SAVE
  • 1-5$110.95 CAD / Pkg of 1
  • 6-29$99.86 CAD / Pkg of 1
  • 30-119$94.31 CAD / Pkg of 1
  • 120+$88.76 CAD / Pkg of 1
Product Description
Key Features
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
More About this Item
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. Autoclavable.
6 Month Warranty
Specifications
  • Standard Size Number13
  • European Size Number70D
  • Top Outside Diameter (OD) (mm)70
  • Bottom Outside Diameter (OD) (mm)60
  • Height (mm)50
  • ColorTranslucent
  • MaterialSilicone
  • DescriptionHigh-purity silicone stopper, size 13 (70D)