Cole-Parmer Griffin Style PP Beaker Starter Set, 5/Pk
Representative image only.
Cole-Parmer Griffin Style PP Beaker Starter Set, 5/Pk
Cole-Parmer | Item: 6310066
Save time and money with beaker starter set
$48.92CAD / Set of 5
BUY MORE & SAVE
  • 1-5$48.92 CAD / Set of 5
  • 6-29$44.03 CAD / Set of 5
  • 30-119$41.58 CAD / Set of 5
  • 120+$39.14 CAD / Set of 5
$48.92CAD / Set of 5
BUY MORE & SAVE
  • 1-5$48.92 CAD / Set of 5
  • 6-29$44.03 CAD / Set of 5
  • 30-119$41.58 CAD / Set of 5
  • 120+$39.14 CAD / Set of 5
Product Description
Key Features
  • Autoclavable
  • 1 year warranty
More About this Item
This polypropylene (PP) starter set includes one 50mL, 100mL, 250mL, 500mL, and 1000mL Griffin style beaker.
The 50- and 100-mL beakers are graduated at 5 mL, the 250- and 500-mL beakers are graduated at 10 mL and the 1000 mL beaker is graduated at 25 mL.
1 Year Warranty
Specifications
  • MaterialPolypropylene(PP)
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)248
  • Withstands Low Temperature (° F)14
  • DescriptionGriffin Style PP Beaker Starter Set, 5/Pk