Cole-Parmer Gallus gallus (Chicken), With Mastermix
Representative image only.
Cole-Parmer Gallus gallus (Chicken), With Mastermix
Cole-Parmer | Item: 9394485
Quickly and accurately verify meat speciation in a food source
Call For Price
Call For Price
Product Description
Includes
Lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
More About this Item
Use the speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.
Specifications
  • Test TypeDNA
  • Number of Tests Included100
  • Mastermix IncludedYes
  • Binomial NameGallus gallus (Chicken)
  • DescriptionGallus gallus (Chicken), With Mastermix