Cole-Parmer Free Chlorine Controller; 110/220 VAC, 50/60 Hz
Representative image only.
Cole-Parmer Free Chlorine Controller; 110/220 VAC, 50/60 Hz
Cole-Parmer | Item: 5610600
Monitor and control drinking water and swimming pool water
$1,760.70CAD / Each
BUY MORE & SAVE
  • 1-5$1,760.70 CAD / Each
  • 6+$1,584.63 CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,760.70CAD / Each
BUY MORE & SAVE
  • 1-5$1,760.70 CAD / Each
  • 6+$1,584.63 CAD / Each
Buy It Together
Current Item
Cole-Parmer Free Chlorine Controller; 110/220 VAC, 50/60 Hz
Cole-Parmer Free Chlorine Controller; 110/220 VAC, 50/60 Hz
5610600
$1,760.70CAD / Each
+ +
Cole-Parmer BNC-to-Stripped Wire Adapter
5610560
$43.17CAD / Each
=
Total Price 2077.55 ADD 3 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Use this chlorine controller for in-line control and measurement of free chlorine ion concentration. This microprocessor-based controller continuously displays the current reading and relay status. Relays are triggered when the concentration reaches high or low setpoints.
A separate alarm relay lets you connect controller to warning lights or buzzers to signal conditions. Use the current output to send data to a recorder or computer for permanent records.
Calibrate this controller from two to five points. Manually activate relays to test pumps, valves, alarms or other process equipment. Security code prevents accidental changes to setup parameters.
1 Year Warranty
Specifications
  • Control TypeOn/Off
  • Display TypeLCD
  • Min pH (pH)0
  • Max pH (pH)20
  • pH Resolution0.001/0.01
  • Output Alarm1 SPDT (220vAC, 5A)
  • Output Current0-20mA or 4-20mA
  • Output Relay2 SPDT relays
  • DescriptionFree Chlorine Controller; 110/220 VAC, 50/60 Hz