Cole-Parmer Folding Skirt Stopper, Silicone, Size No. 19.4; 20/PK
Representative image only.
Cole-Parmer Folding Skirt Stopper, Silicone, Size No. 19.4; 20/PK
Cole-Parmer | Item: 6299595
Stay watertight even after several perforations
$191.17CAD / Pkg of 20
BUY MORE & SAVE
  • 1-5$191.17 CAD / Pkg of 20
  • 6-29$172.05 CAD / Pkg of 20
  • 30-119$162.49 CAD / Pkg of 20
  • 120+$152.94 CAD / Pkg of 20
$191.17CAD / Pkg of 20
BUY MORE & SAVE
  • 1-5$191.17 CAD / Pkg of 20
  • 6-29$172.05 CAD / Pkg of 20
  • 30-119$162.49 CAD / Pkg of 20
  • 120+$152.94 CAD / Pkg of 20
Product Description
Key Features
  • Easily extract contents by perforating stopper
  • 6 month warranty
More About this Item
Two-part folding skirt stoppers are designed to keep your contents secure two ways. The lower section is molded to grip the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim. Ideal for securing glass, metal, and plastic containers regardless of type. Upper hollow plug section can be perforated to exact contents without removing the stopper — remains watertight even after several perforations.
The rubber stoppers are ideal for chemical transfer and withstand temperatures from -4°F (-20°C) to 212°F (100°C). Chemical-resistent silicone stoppers are recommended for biological applications and withstand temperatures from -76°F (-60°C) to 428°F (220°C).
6 Month Warranty
Specifications
  • Standard Size Number19.4
  • Top Outside Diameter (OD) (mm)17.8
  • Bottom Outside Diameter (OD) (mm)15.8
  • Height (mm)32.5
  • ColorNatural
  • MaterialSilicone
  • DescriptionFolding Skirt Stopper, Silicone, Size No. 19.4; 20/PK